Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD47 Mouse anti-Human, Alexa Fluor™ 488, Clone: CC2C6, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568339
This item is not returnable.
View return policy
Description
CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for Flow cytometry (Routinely Tested)Specifications
CD47 | |
Monoclonal | |
Alexa Fluor 488 | |
CD47; IAP; integrin associated protein; MER6; OA3; Rh-related antigen | |
Human CCRF-CEM | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG1 |
Flow Cytometry | |
CC2C6 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title