Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD47 Rabbit anti-Human, Clone: CD47/6362R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen CD47
Clone CD47/6362R
Concentration 0.2 mg/ml
Dilution ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121921
View Documents
Novus Biologicals
NBP30722410UG
100 μg
Each for $406.52
Add to cart
 
NB121922
View Documents
Novus Biologicals
NBP30722420UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

CD47
0.2 mg/ml
ELISA, Immunohistochemistry (Paraffin)
Purified
RUO
Human
antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen
A recombinant fragment (around aa 18-135) of human CD47 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
CD47/6362R
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Immunology, Signal Transduction
10 mM PBS with 0.05% BSA
961
IgG Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit