Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD48 Monoclonal Antibody (OX-45), PE
Mouse Monoclonal Antibody
$319.00 - $634.50
Specifications
Antigen | CD48 |
---|---|
Clone | OX-45 |
Concentration | 0.1 mg/mL |
Content And Storage | 4°C, do not freeze |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA517528
|
Invitrogen™
MA517528 |
50 μg |
Each for $319.00
|
|
|||||
PIMA517529
|
Invitrogen™
MA517529 |
200 μg |
Each for $634.50
|
|
|||||
Description
CD48 Monoclonal Antibody for Flow
CD48 (Blast-1) belongs to the CD2 subset of the Ig superfamily that includes CD2, CD2F-10, CD58, CD84, CD150, CD229, CD244 and others. These molecules bind to the same or another members of the Ig family, thus mediating homotypic or heterotypic adhesion. CD48 is a GPI-anchored protein broadly expressed on hematopoietic cells and serves as a high affinity ligand for 2B4 and low affinity ligand for CD2. 2B4-CD48 interaction among NK cells and NK-T cells regulates cell proliferation. Further, signaling through CD48 results in eosinophil activation and CD48 expression is increased in several infectious diseases. CD48 is strongly expressed on lymphocytes and monocytes, weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of marker for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2. 2B4-CD48 interaction may be involved in the pathology of allergic airway diseases and provide a pharmacological target for the treatment of diseases such as allergic rhinitis and asthma.Specifications
CD48 | |
0.1 mg/mL | |
Flow Cytometry | |
PE | |
Mouse | |
RUO | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P10252 | |
245962 | |
rat T cell blasts (stimulated purified T helper cells with allogeneic irradiated rat spleen cells). | |
Antibody |
OX-45 | |
4°C, do not freeze | |
Monoclonal | |
Liquid | |
IgG1 | |
Rat | |
Cd48 | |
AI449234; AW610730; B-cell activation marker; BCM1; Bcm-1; BCM1 surface antigen; BLAST; BLAST1; BLAST-1; B-lymphocyte activation marker BLAST-1; Cd48; CD48 antigen; CD48 antigen (B cell membrane protein); CD48 antigen (B-cell membrane protein); Cd48 molecule; CD48d; hCD48; HM48-1; leukocyte antigen MEM-102; mCD48; MEM-102; MRC OX-45 surface antigen; sCD 48; sCD48; Sgp-60; Signaling lymphocytic activation molecule 2; Signaling lymphocytic activation molecule 2 (SLAMF2); SLAM family member 2; SLAMF2; soluble CD 48; soluble CD48; TCT.1 | |
Cd48 | |
Primary | |
Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title