Learn More
CD48/SLAMF2 Mouse anti-Human, Alexa Fluor 700, Clone: 156-4H9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143403AF700
Catalog No. NBP143403AI
Add to cart
Description
Description
CD48/SLAMF2 Monoclonal specifically detects CD48/SLAMF2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD48/SLAMF2
156-4H9
Flow Cytometry
Purified
CD48
The immunogen for this antibody was CD48.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Flow Cytometry
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2
Mouse
IgG1
0.1 ml
Cell Biology, Cellular Markers, Immunology, Stem Cell Markers
Primary
962