Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD48/SLAMF2 Rabbit anti-Rat, Alexa Fluor 488, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289667AF488

Catalog No. NB067623


Add to cart

Description

Description

CD48/SLAMF2 Monoclonal antibody specifically detects CD48/SLAMF2 in Rat samples. It is validated for Flow Cytometry
Specifications

Specifications

CD48/SLAMF2
2
Flow Cytometry
Purified
Rabbit
IgG
0.1 mL
Cell Biology, Cellular Markers, Immunology, Stem Cell Markers
Primary
962
Flow Cytometry
Alexa Fluor 488
50mM Sodium Borate
BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2
Recombinant Rat CD48/SLAMF2 Protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit