Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD49e (Integrin alpha 5) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA586819
Description
Integrin alpha-5, also known as FNRA or VLA5A, is a protein that in humans is encoded by the ITGA5 gene. The product of this gene belongs to the integrin alpha chain family. Integrins are integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signaling.Specifications
CD49e (Integrin alpha 5) | |
Polyclonal | |
Unconjugated | |
ITGA5 | |
Integrin alpha-5; CD49 antigen-like family member E; Fibronectin receptor subunit alpha; Integrin alpha-F; VLA-5; CD49e; ITGA5; FNRA | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16402, 315346, 3678 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P08648, P11688 | |
Itga5 | |
A synthetic peptide corresponding to residues in Human ITGA5 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title