Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITGA5 Rat anti-Mouse, Biotin, Clone: 5H10.27 (MFR5), Invitrogen™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen A15402

Catalog No. 50-113-7027


Add to cart

Description

Description

The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
Specifications

Specifications

ITGA5
Monoclonal
0.5 mg/mL
PBS with 0.1% sodium azide
P11688
FNRA, CD49e, VLA5A, ITGA5
Rat
IgG2a, kappa
100 μg
4° C
Primary
16402
Flow Cytometry
5H10.27 (MFR5)
Biotin
Itga5
Liquid
Itga5
C57BL/6 x A/J F1 mouse mast cell line MC/9 1
Purified
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit