Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD49e Rat anti-Mouse, BV786, Clone: 5H10-27 (MFR5), BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740863
Description
CD49e Monoclonal antibody specifically detects CD49e in Mouse samples. It is validated for Flow Cytometry.Specifications
CD49e | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
5H10-27 (MFR5) | |
Brilliant Violet 786 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title