Learn More
Epredia™ Lab Vision™ CD5 Ab-1, Mouse Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ CD5 Ab-1, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS393S0
Description
CD5, a transmembrane protein, is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphoma, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas (same as diffuse centrocytic lymphomas) are CD5+ while the follicle center cell lymphoma are CD5-.
Host Species: Mouse
Clone: 4C7
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: External domain
Immunogen: Recombinant protein corresponding to the external domain of the CD5 molecule
Molecular Weight: 67kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Specifications
CD5 Ab-1 | |
Monoclonal | |
Unconjugated | |
Recombinant protein corresponding to the external domain of the CD5 molecule | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
4C7 | |
Mouse | |
100 μL | |
Hematopoietic | |
Human | |
IgG2a κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.