Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CD5 Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2795765PLATES

 View more versions of this product

Catalog No. NB010192


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Mouse CD5.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
12.5 pg/mL
921
RUO
CD5
Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology, Phospho Specific
Mouse
CD5
1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 800 pg/mL is recommended.
HRP
This solid phase sandwich ELISA utilizes an antibody specific for Mouse CD5.
Sandwich ELISA
  • Standard: Each vial contains 140 ng of recombinant mouse CD5.
  • Capture Antibody: 1 mg/mL of rabbit anti-mouse CD5 monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL rabbit anti-CD5 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Storage is content dependent.
    Rabbit
    5 Plates
    12.5-800 pg/mL
    CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit