Learn More
CD5 Mouse anti-Human, Clone: OTI6C9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802406
Catalog No. 50-166-5990
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter ft.s Syndrome.
Specifications
Specifications
CD5
Monoclonal
Unconjugated
CD5
Lyophilized
CD5
Full length human recombit protein of human CD5 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
OTI6C9
PBS with 8% trehalose and no preservative; pH 7.3
P06127
LEU1, T1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
921