Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD5 Mouse anti-Human, PE, Clone: OTI7A7, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270360PE
Description
CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Flow Cytometry.Specifications
CD5 | |
OTI7A7 | |
Flow Cytometry | |
Affinity Purified, Conjugated | |
CD5 | |
Full length recombinant protein of human CD5 (NP_055022) produced in HEK293T cells. | |
Immunogen affinity purified | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology, Phospho Specific | |
Primary | |
921.0 |
Flow Cytometry | |
PE | |
PBS with 0.05% Sodium Azide | |
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title