Learn More
CD5 Mouse anti-Human, PE-Cyanine5.5, Clone: CD5-5D7, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD0518
Catalog No. 50-112-1317
Add to cart
Description
Description
The R-phycoerythrin (PE)-Cy5.5™ tandem conjugate permits simultaneous multicolor labeling and detection of multiple targets with excitation by a single excitation source-the 488 nm line of the argon-ion laser.
CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter’s Syndrome.
Specifications
Specifications
CD5
Monoclonal
PE-Cyanine5.5
CD5
Liquid
CD5
Human CD5
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
CD5-5D7
PBS with 0.1% sodium azide
P06127
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
921