Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5 Mouse anti-Human, PE-Cyanine7, Clone: OTI7A7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270360PECY7

Catalog No. N270360PEC7


Add to cart

Description

Description

CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD5
OTI7A7
Flow Cytometry
Affinity Purified, Conjugated
CD5
Full length recombinant protein of human CD5 (NP_055022) produced in HEK293T cells.
Immunogen affinity purified
Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology, Phospho Specific
Primary
921.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit