Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5 Mouse anti-Human, PerCP-Cyanine5.5, Clone: L17F12, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

$358.00

Specifications

Antigen CD5
Clone L17F12
Target Species Human
Conjugate PerCP-Cyanine5.5
Isotype IgG2a, kappa
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-112-3697 Non-distribution product as customer accommodation. View Documents Invitrogen
45005842
100 Tests Each for $358.00
Add to cart
 
Description

Description

Description: The L17F12 monoclonal antibody reacts with human CD5, a 67 kDa protein expressed by a majority of thymocytes and mature T cells and a subset of B cells. Signaling through the CD5 molecule activates T cells and binding of CD5 to its ligand on B cells, CD72, and plays an important role in T-B interaction and proliferation. Applications Reported: This L17F12 antibody has been reported for use in flow cytometric analysis. Applications Tested: This L17F12 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.06 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser. Filtration: 0.2 µm post-manufacturing filtered.

CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter’s Syndrome.
Specifications

Specifications

CD5
Human
IgG2a, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
L17F12
PerCP-Cyanine5.5
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit