Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD5 Rat anti-Murine, Brilliant Ultraviolet 737, Clone: 53-7.3, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 612809
Description
CD5 Monoclonal antibody specifically detects CD5 in Mouse samples. It is validated for Flow cytometry.Specifications
CD5 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Purified | |
RUO | |
Mouse |
Flow Cytometry | |
53-7.3 | |
Brilliant Ultraviolet 737 | |
Lymphocyte antigen 1; Cd5; Ly-12; Ly-A; Lyt-1 | |
Mouse Thymus / Spleen | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title