Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD51 Mouse anti-Human, Alexa Fluor 647, Clone: NKI-M9, BD Horizon™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 565835

Catalog No. BDB565835


Add to cart

Description

Description

CD51 Monoclonal antibody specifically detects CD51 in Human samples. It is validated for Flow cytometry.
Specifications

Specifications

CD51
NKI-M9
Alexa Fluor 647
Purified
Murine
Mouse BALB/c IgG2a, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Integrin alpha-V; alphaV; αv; ITGAV; MSK8; VNRA; VTNR
Human Melanoma Cells
25μg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Reseach Use Only.