Learn More
CD51 Rat anti-Mouse, BV786, Clone: RMV-7, BD OptiBuild™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 740946
Catalog No. BDB740946
Add to cart
Description
Description
CD51 Monoclonal antibody specifically detects CD51 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD51
RMV-7
Brilliant Violet 786
Rat
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG1, κ
RUO
Primary
Mouse