Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD52 Antibody (Hu116), HRP, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals FAB9889H
Description
CD52 Monoclonal antibody specifically detects CD52 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| CD52 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Human | |
| 0.1 mL | |
| Cancer, Immunology | |
| 1043 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| Hu116 | |
| Flow Cytometry, CyTOF-ready | |
| Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5 | |
| Human CD52 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction