Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD52 Chimeric Recombinant Rabbit Monoclonal Antibody (YTH 34.5-G2b (Campath-1G)), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA547807

Encompass

Catalog No. PIMA547807


Add to Cart

Description

Description

Specificity: Human CD52 antigen, also known as CAMPATH-1. CD52 is expressed at high density by lymphocytes, monocytes, eosinophils, thymocytes and macrophages. It is expressed by most lymphoid derived malignancies, although expression on myeloma cells is variable.

CD52 Chimeric Recombinant Monoclonal antibody specifically detects CD52 Chimeric in Human samples. It is validated for Cytotoxicity Assay, ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD52 Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
P06729
Rabbit
Affinity chromatography
RUO
914
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Cytotoxicity Assay, ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen)
YTH 34.5-G2b (Campath-1G)
Unconjugated
CD2
AI463198; B7; B7-Ag; cambridge pathology 1 antigen; CAMPATH-1; CAMPATH-1 antigen; Cd52; CD52 antigen; CD52 antigen (CAMPATH-1 antigen); CD52 molecule; CDW52; CDW52 antigen (CAMPATH-1 antigen); CLS1; Epididymal secretory protein E5; epididymis secretory sperm binding protein Li 171mP; he5; HEL-S-171mP; human epididymis-specific protein 5; Lymphocyte differentiation antigen B7; Mb7
Proprietary sequence.
200 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit