Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD52 Human anti-Human, Cynomolgus Monkey, Rhesus Macaque, DyLight™ 350, Clone: Campath-1H, Novus Biologicals™

Human Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275896UV

Catalog No. NBP275896UV


Add to cart

Description

Description

CD52 Monoclonal antibody specifically detects CD52 in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence.
Specifications

Specifications

CD52
Campath-1H
ELISA, Immunohistochemistry, Immunofluorescence
Conjugated, Purified
CD52
Human lymphocytes
Protein A purified
Store at 4°C in the dark.
Monoclonal
Human CD52 antigen, also known as CAMPATH-1. CD52 is expressed at high density by lymphocytes, monocytes, eosinophils, thymocytes and macrophages. It is expressed by most lymphoid derived malignancies, although expression on myeloma cells is variable.
ELISA, Immunofluorescence, Immunohistochemistry
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5
Human
IgG1 Kappa
0.1 mL
Primary
1043.0
Cynomolgus Monkey, Human, Rhesus Macaque
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit