Learn More
CD52 Human anti-Human, PE, Clone: Hu116, R&D Systems™
Human Monoclonal Antibody
Manufacturer:  R&D Systems FAB9889P100
Catalog No. FAB9889P100
Add to cart
This item is not returnable. View return policy
Description
Description
CD52 Monoclonal antibody specifically detects CD52 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
CD52
Hu116
Flow Cytometry 10 uL/10^6 cells
Purified
Human
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Human
Flow Cytometry
PE
Supplied in a saline solution containing BSA and Sodium Azide.
Cambridge pathology 1 antigen, CAMPATH-1, CAMPATH-1 antigen, CD52, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, HEL-S-171mP, Human epididymis-specific protein 5
Human CD52
Protein A or G purified from cell culture supernatant
RUO
Primary
1043