Learn More
CD52 Mouse anti-Human, APC, Clone: CF1D12, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD5205
Catalog No. 50-112-1435
Add to cart
Description
Description
Allophycocyanin (APC) is a stable and highly soluble phycobiliprotein that provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing exceptional quantum yields and molar extinction coefficients.
Human CD52 is a glycosylphosphatidylinositol (GPI) anchored antigen expression in a number of different tissues.
Specifications
Specifications
CD52
Monoclonal
APC
CD52
Liquid
CD52
Human CD52
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
CF1D12
PBS with 10% sucrose, BSA and 0.1% sodium azide
P31358
CDW52, CAMPATH-1 antigen, CD52 antigen, cambridge pathology 1 antigen, epididymal secretory protein E5, he5, human epididymis-specific protein 5
Mouse
IgG3
0.5 mL
4° C, store in dark
Primary
1043