Learn More
CD52 Rabbit anti-Human, Monkey, Alexa Fluor™ 532, Clone: YTH 34.5-G2b (Campath-1G), Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252657AF532
Catalog No. NP2267A32
Add to cart
Description
Description
CD52 Monoclonal antibody specifically detects CD52 in Human,Monkey samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
CD52
YTH 34.5-G2b (Campath-1G)
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
Human CD52 antigen, also known as CAMPATH-1. CD52 is expressed at high density by lymphocytes, monocytes, eosinophils, thymocytes and macrophages. It is expressed by most lymphoid derived malignancies, although expression on myeloma cells is variable.
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CD52
Human peripheral mononuclear cells enriched for T-cells.
0.1 mL
Primary
1043
Human, Monkey