Learn More
CD53 Mouse anti-Human, FITC, Clone: MEM-53, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15736
Catalog No. 50-113-7085
Add to cart
Description
Description
CD53 is a tetraspanin family transmembrane glycoprotein expressed in the lymphoid-myeloid lineage. This molecule has been reported to form complexes with other leukocyte surface proteins such as CD2, CD19, CD21, MHC II, VLA-4 or tetraspanins CD37, CD81 and CD82, thus probably modulating various signaling processes. CD53 is involved in radioresistancy of tumour cells and its triggering has anti-apoptotic effect. In thymus, CD53 is up-regulated in response to positive selection signals during T cell development, and is strongly expressed upon macrophage exposure to bacterial lipopolysaccharide, whereas stimulation of neutrophils results in down-regulation of CD53 expression.
Specifications
Specifications
CD53, CD53
Monoclonal
FITC
CD53
Liquid
CD53
Human CD53
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MEM-53
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P19397
MOX44, TSPAN25, CD53 antigen, CD53 glycoprotein, CD53 tetraspan antigen
Mouse
IgG1
100 tests
4° C
Primary
963