Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD53 Rat anti-Mouse, BB515, Clone: OX-79, BD Horizon™

Rat Monoclonal Antibody

$112.40 - $176.80

Specifications

Antigen CD53
Isotype Rat AO IgM κ
Immunogen BAB/14 mouse macrophage cell line RAW 264
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Conjugate BB515
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566061
View Documents Promotion Details
BD Biosciences
566061
25 μg
Each for $112.40
Add to cart
 
BDB565288
View Documents Promotion Details
BD Biosciences
565288
50 μg
Each for $176.80
Add to cart
 
Description

Description

CD53 Monoclonal antibody specifically detects CD53 in Mouse samples. It is validated for Flow cytometry.

CD53 Monoclonal antibody specifically detects CD53 in Mouse samples. It is validated for Flow cytometry.
Specifications

Specifications

CD53
BAB/14 mouse macrophage cell line RAW 264
BB515
Cell Biology
Rat
Purified
0.2mg/mL
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Rat AO IgM κ
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Flow Cytometry
RUO
OX-79
Cd53: Ox-44; Tspan25; Tetraspanin-25
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit