Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD54 Mouse Anti-Rat, Brilliant Violet 421, Clone: 1A29, BD Optibuild™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 742404

Catalog No. BDB742404


Add to cart

Description

Description

The 1A29 monoclonal antibody specifically recognizes ICAM-1 which is also known as CD54. ICAM-1 is an 85 kDa type I transmembrane glycoprotein that is encoded by Icam1 (Intercellular adhesion molecule 1). It is expressed on vascular endothelium in lymphoid tissues, thymic stromal cells, peripheral blood monocytes, peritoneal macrophages and mast cells, dendritic cells, and weakly on peripheral lymphocytes and thymocytes. ICAM-1 is a ligand for LFA-1 (CD11a/CD18) and Mac-1 (CD11b/CD18). Its expression is upregulated on activated lymphocytes and endothelial cells. 1A29 mAb inhibits Phorbol 12-Myristate 13-Acetate (PMA)-induced aggregation of phytohemagglutinin (PHA)-stimulated splenic blasts, as well as the adhesion of mitogen-stimulated blasts to high endothelial venule (HEV) cells and purified ICAM-1. The 1A29 antibody can reportedly inhibit leucocyte infiltration in in vivo systems, blocks induction of experimental allergic encephalomyelitis, and reduces NK-cell adhesion to tumor cells.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications

Specifications

CD54
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
ICAM-1, Icam1, ICAM, Intercellular adhesion molecule 1
Rat HEV-Derived Cell Line Ax
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV421 under optimal conditions that minimize unconjugated dye and antibody.
RUO
Primary
Murine
1A29
Brilliant Violet 421
Affinity Purified
Murine
Mouse BALB/c IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit