Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD54 Mouse Anti-Rat, Brilliant Violet 650, Clone: 1A29, BD Optibuild™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 742407

Catalog No. BDB742407


Add to cart

Description

Description

The 1A29 monoclonal antibody specifically recognizes ICAM-1 which is also known as CD54. ICAM-1 is an 85 kDa type I transmembrane glycoprotein that is encoded by Icam1 (Intercellular adhesion molecule 1). It is expressed on vascular endothelium in lymphoid tissues, thymic stromal cells, peripheral blood monocytes, peritoneal macrophages and mast cells, dendritic cells, and weakly on peripheral lymphocytes and thymocytes. ICAM-1 is a ligand for LFA-1 (CD11a/CD18) and Mac-1 (CD11b/CD18). Its expression is upregulated on activated lymphocytes and endothelial cells. 1A29 mAb inhibits Phorbol 12-Myristate 13-Acetate (PMA)-induced aggregation of phytohemagglutinin (PHA)-stimulated splenic blasts, as well as the adhesion of mitogen-stimulated blasts to high endothelial venule (HEV) cells and purified ICAM-1. The 1A29 antibody can reportedly inhibit leucocyte infiltration in in vivo systems, blocks induction of experimental allergic encephalomyelitis, and reduces NK-cell adhesion to tumor cells.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
Specifications

Specifications

CD54
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
ICAM-1, Icam1, ICAM, Intercellular adhesion molecule 1
Rat HEV-Derived Cell Line Ax
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV650 under optimal conditions that minimize unconjugated dye and antibody.
RUO
Primary
Murine
1A29
Brilliant Violet 650
Affinity Purified
Murine
Mouse BALB/c IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit