Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CD55/DAF Antibody Pair [HRP]

Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2794405PLATES

 View more versions of this product

Catalog No. NB009920


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD55/DAF.
Specifications

Specifications

HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD55/DAF.
23.44 pg/mL
1604
RUO
CD55/DAF
Immunology
Human
CD55
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.2 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 1.5 ng/mL is recommended
Sandwich ELISA
Antibody Pairs
Sandwich ELISA
  • Standard: Each vial contains 30 ng of recombinant CD55/DAF.
  • Capture Antibody: 1.0 mg/mL of rabbit anti-CD55/DAF monoclonal antibody.
  • Detection Antibody: 0.4 mg/mL mouse anti-CD55/DAF monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Storage is content dependent.
    Mouse/Rabbit
    5 Plates
    23.44-1500 pg/mL
    CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit