Learn More
CD55/DAF Mouse anti-Human, Janelia Fluor 646, Clone: 67, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP250226JF646
Catalog No. NBP50226646
Add to cart
Description
Description
CD55/DAF Monoclonal antibody specifically detects CD55/DAF in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
CD55/DAF
67
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
CD55
Fibronectin purified human monocytes .
Protein G purified
Immunology
Primary
1604
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
Human