Learn More
CD55/DAF Rat anti-Mouse, Alexa Fluor 700, Clone: 583905, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB5376N100UG
Catalog No. FAB5376N100
Add to cart
This item is not returnable. View return policy
Description
Description
CD55/DAF Monoclonal specifically detects CD55/DAF in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD55/DAF
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q61475
CD55
Mouse myeloma cell line NS0-derived recombinant mouse CD55/DAF Met1-Pro359 Accession # Q61475
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse CD55/DAF in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CD55 or recombinant mouse CD97 is observed.
Flow Cytometry
583905
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD55, CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CR, CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, DAF, decay accelerating factor for complement, TC
Rat
IgG2a
RUO
Primary
1604
Mouse
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)