Learn More
CD55 Mouse anti-Human, BB515, Clone: IA10, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565939
Catalog No. BDB565939
Add to cart
Description
Description
CD55 Monoclonal antibody specifically detects CD55 in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
CD55
IA10
Aqueous buffered solution containing ≤0.09% sodium azide.
DAF; CR; CROM; TC; Complement decay-accelerating factor
Mouse IgG2a, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Monoclonal
Flow Cytometry
BB515
Purified
Mouse
25 Tests
Cell Biology
Primary
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.