Learn More
CD55 Mouse, anti-Human, R718, Clone: IA10, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 752109
Catalog No. BDB752109
Add to cart
Description
Description
The IA10 monoclonal antibody specifically binds to CD55, which is also known as complement decay-accelerating factor (DAF). CD55 is a glycophosphatidylinositol (GPI)-anchored, single chain membrane glycoprotein of approximately 70 kDa that belongs to the regulators of complement activation (RCA) gene family which includes CD21, CD35, and CD46. CD55 is widely expressed on hematopoietic cells including platelets and erythrocytes, as well as on many non-hematopoietic cells, such as, endothelial cells and epithelial cells. CD55 is involved in protecting cells from damage by autologous activated complement complexes. CD55 prevents the amplification steps of the complement cascade by interfering with the assembly of the C3-convertases, C4b2a and C3bBb, and the C5-convertases, C4b2a3b and C3bBb3b.
Specifications
Specifications
CD55
IA10
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Human