Learn More
CD55 Mouse anti-Human, Unconjugated, Clone: 143-30, Abnova™
Mouse Monoclonal Antibody.
Supplier:  Abnova Corporation MAB20907
Catalog No. 89-337-402
Add to Cart
Description
Description
CD55 Monoclonal antibody specifically detects CD55 in Human samples. It is validated for IHC-Fr, IF, Flow Cyt.
Specifications
Specifications
CD55
Monoclonal
Unconjugated
CD55
Mouse
100 μg
1604
Store at 2 to 8°C
IgG1 κ
Immunohistochemistry (Frozen), Immunofluorescence, Flow Cytometry
143-30
CD55 molecule, decay accelerating factor for complement (Cromer blood group)
CR, CROM, DAF, TC
PHA stimulated human PBL
Primary
Human
Liquid
Product Suggestions
Product Suggestions