Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD56 Mouse anti-Human , BB700, Clone: NCAM16.2 (also known as NCAM 16), BD Biosciences™

Mouse Monoclonal Antibody

$193.70 - $426.50

Specifications

Antigen CD56
Clone NCAM16.2 (also known as NCAM 16)
Applications Flow Cytometry
Conjugate BB700
Format Conjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566574
View Documents
BD Biosciences
566574
25 Tests
Each for $193.70
Add to cart
 
BDB566573
View Documents
BD Biosciences
566573
100 Tests
Each for $426.50
Add to cart
 
Description

Description

CD56 Monoclonal antibody specifically detects CD56 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD56
Flow Cytometry
Conjugated
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
Immunoaffinity-enriched adult human brain NCAM
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB700 under optimum conditions, and unconjugated antibody and free BD Horizon BB700 were removed.
NCAM16.2 (also known as NCAM 16)
BB700
Mouse
Human
NCAM1; NCAM-1; NCAM; Leu-19; Neural cell adhesion molecule 1; NKH1; MSK39
Mouse BALB/c IgG2b, κ
Primary
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit