Learn More
CD56 Mouse anti-Human, PE-Texas Red®, Clone: MEM-188, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD5617
Catalog No. 50-112-1442
Add to cart
Description
Description
The R-phycoerythrin (PE)-Texas Red™ conjugate permits simultaneous multicolor labeling and detection of multiple targets with excitation by a single excitation source-the 488 nm line of the argon-ion laser. This antibody recognizes an isoform of neural cell adhesion molecule (NCAM) also known as NKH-1.
NCAM, as a member of the immunoglobulin superfamily of adhesion molecules is characterized by several immunoglobulin (Ig)-like domains. The extracellular part of NCAM consists of five of these Ig domains and two fibronectin type III homology regions. NCAM is encoded by a single copy gene composed of 26 exons. However, at least 20-30 distinct isoforms can be generated by alternative splicing and by posttranslational modifications, such as sialylation. During sialylation, polysialic acid (PSA) carbohydrates are attached to the extracellular part of NCAM. Through its extracellular region, NCAM mediates homophilic interactions. In addition, NCAM can also undergo heterophilic interactions by binding extracellular matrix components, such as laminin, or other cell adhesion molecules, such as integrins.
Specifications
Specifications
CD56
Monoclonal
PE-Texas Red™
NCAM1
Liquid
NCAM1
Human CD56
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
MEM-188
PBS with BSA, sucrose and 0.1% sodium azide
P13591
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Mouse
IgG2a, kappa
0.5 mL
4° C, store in dark
Primary
4684