Learn More
Novus Biologicals™ CD59 Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
Manufacturer:  Novus Biologicals™ NBP2794015PLATES
 View more versions of this product
Catalog No. NB009842
Add to cart
Description
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD59.
Specifications
Specifications
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD59.
234.38 pg/mL
966
RUO
CD59
Cell Biology, Cellular Markers, Immunology, Signal Transduction, Stem Cell Markers
Human
CD59
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 15000 pg/mL is recommended.
Antibody Pairs
Sandwich ELISA
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human CD59 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.5 mg/mL of rabbit anti-Human CD59 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 240 ng of recombinant Human CD59.
  • Storage is content dependent.
    Rabbit
    5 Plates
    234.38-15000 pg/mL
    16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein