Learn More
CD59 Mouse anti-Human, Clone: p282 (H19), Abnova™
Mouse monoclonal antibody raised against human CD59.
Manufacturer:  Abnova Corporation MAB12534
Catalog No. 89-933-847
Add to cart
Description
Description
This gene encodes a cell surface glycoprotein that regulates complement-mediated cell lysis, and it is involved in lymphocyte signal transduction. This protein is a potent inhibitor of the complement membrane attack complex, whereby it binds complement C8 and/or C9 during the assembly of this complex, thereby inhibiting the incorporation of multiple copies of C9 into the complex, which is necessary for osmolytic pore formation. This protein also plays a role in signal transduction pathways in the activation of T cells. Mutations in this gene cause CD59 deficiency, a disease resulting in hemolytic anemia and thrombosis, and which causes cerebral infarction. Multiple alternatively spliced transcript variants, which encode the same protein, have been identified for this gene. [provided by RefSeq]
Specifications
Specifications
CD59
p282 (H19)
Mouse monoclonal antibody raised against human CD59.
In PBS, pH 7.2 (0.09% sodium azide).
16.3A5/1F5/EJ16/EJ30/EL32/FLJ38134/FLJ92039/G344/HRF-20/HRF20/MAC-IP/MACIF/MEM43/MGC2354/MIC11/MIN1/MIN2/MIN3/MIRL/MSK21/p18-20
Mouse
IgG2a, kappa
50 ug
Store at 4°C. Do not freeze.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (2 ug/106 cells) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
CD59
Human CD59
Affinity purification
RUO
Primary
966