Learn More
CD59 Mouse anti-Human, HRP, Clone: 8D2B8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237436H
Catalog No. NBP237436H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD59
8D2B8
Western Blot, ELISA, Immunohistochemistry
Purified
CD59
Purified recombinant fragment of human CD59 (AA: 31-111) expressed in E. Coli.
Protein A or G purified
Cell Biology, Cellular Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary
966
ELISA, Immunohistochemistry, Western Blot
HRP
PBS with No Preservative
16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human