Learn More
Abnova™ CD6 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000923T01
Catalog No. 89-938-748
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CD6 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 68.9kDa
Transient overexpression cell lysate was tested with anti-CD6 antibody (Mfr. No. H00000923-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
BC033755.1
pCMV-CD6 full-length
100 μL
Human