Learn More
CD6 Mouse anti-Human, Clone: UMCD6/3F7B5, Abnova™
Mouse monoclonal antibody raised against human CD6.
Manufacturer:  Abnova Corporation MAB12898
Catalog No. 89-933-852
Add to cart
Description
Description
CD6 is a monomeric 105- or 130-kD membrane glycoprotein that is involved in T-cell activation. The size difference between the 2 CD6 forms is due to differences in phosphorylation (Robinson et al., 1995 [PubMed 7589069]).[supplied by OMIM]
Specifications
Specifications
CD6
UMCD6/3F7B5
Mouse monoclonal antibody raised against human CD6.
In 50mM Na
FLJ44171/TP120
Mouse
IgG1, kappa
50 ug
Store at 4°C. Do not freeze.
Monoclonal
Human
Flow Cytometry, Functional Assay, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry Functional Study Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
CD6
Human rheumatoid synovial T cell line (ST-1).
Protein A purification
RUO
Primary
923