Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD6 Rabbit anti-Human, Unconjugated, Clone: C6/2884R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00667

Catalog No. 89-339-682


Add to cart

Description

Description

CD6 Monoclonal antibody specifically detects CD6 in Human samples. It is validated for IHC-P, IF, Flow Cyt.
Specifications

Specifications

CD6
C6/2884R
CD6 molecule
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human
Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Unconjugated
CD6
FLJ44171, TP120
Recombinant protein corresponding to full-length human CD6
100 μg
Primary
923
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit