Learn More
CD6 Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0190-2F1, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212173AF594
Catalog No. NP212173A94
Add to cart
Description
Description
CD6 Monoclonal antibody specifically detects CD6 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CD6
RM0190-2F1
Western Blot
CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CD6
Mouse recombinant protein CD6
0.1 mL
Primary
923