Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Lab Vision™ CD61 (GPIIIa) Ab-1, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD61 (GPIIIa) Ab-1, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS1095R1
Description
CD61 (GPIIIa) is a glycoprotein found on megakaryocytes, platelets and their precursors. CD61 antigen plays a role in platelet aggregation and also as a receptor for fibrinogen, fibronectin, von Willebrand factor and vitronectrin.Host Species: Mouse
Clone: 2f2
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Extracellular domain
Immunogen: Recombinant protein encoding part of the external domain of human CD61
Molecular Weight: 105 kDa
Positive Control: Tonsil
Cellular Localization: Membrane of platelets
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
CD61 (GPIIIa) Ab-1 | |
Monoclonal | |
Unconjugated | |
Recombinant protein encoding part of the external domain of human CD61 | |
RUO | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
2f2 | |
Mouse | |
1 mL | |
Hematopoietic | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction