Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD61 Monoclonal Antibody (HCD61-AP3), Abbomax
Mouse Monoclonal Antibody
Supplier: Abbomax Inc 603800
Description
Positive control: Human peripheral blood platelets Cellular location: Membrane, Cytoplasmic.
CD61 Monoclonal antibody specifically detects CD61 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)Specifications
CD61 | |
Monoclonal | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P05106 | |
Mouse | |
Protein A | |
RUO | |
3690 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
HCD61-AP3 | |
Unconjugated | |
ITGB3 | |
antigen CD61; BDPLT16; BDPLT2; beta3 integrin; CD61; EGK_08849; glycoprotein GPIIIa; glycoprotein IIIa; GP3A; GPIIIa; GT; INGRB3; integrin beta 3; integrin beta 3 (Cd61); integrin beta-3; integrin beta-3 subunit; integrin subunit beta 3; integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGB3; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet gpIIIa; platelet membrane glycoprotein IIIa | |
Mice were immunized with human platelets | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction