Learn More
CD62L Monoclonal Antibody (1H3), Exalpha

Mouse Monoclonal Antibody
Supplier: Vector Laboratories 0621100UG


Description
Flow Cytometry PBMC: Add 10 μL of MAB/10^6 PBMC in 100 μL PBS. Mix gently and incubate for 15 minutes at 2 to 8C. Wash twice with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. Flow Cytometry Whole Blood: Add10 μL of MAB/100 μL of whole blood. Mix gently and incubate for 15 minutes at room temperature 20C. Lyse the whole blood. Wash once with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. See instrument manufacturer instructions for Lysed Whole Blood and Immunofluorescence analysis with a flow cytometer or microscope.
CD62L Monoclonal antibody specifically detects CD62L in Human samples. It is validated for Flow CytometrySpecifications
CD62L | |
Monoclonal | |
Unconjugated | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P14151 | |
Mouse | |
Protein A/G | |
RUO | |
6402 | |
4° C, do not freeze | |
Liquid |
Flow Cytometry | |
1H3 | |
100UG HU CD11B C67F154 AFP647 MAB 100 UG | |
SELL | |
A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1 | |
CD62L was derived from the hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immunized with peripheral blood T lymphocytes. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.