Learn More
CD62L Mouse anti-Human, FITC, Clone: Dreg-56, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD62L01
Catalog No. 50-112-1447
Add to cart
Description
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD62L (L-selectin) is an adhesion glycoprotein that is constitutively expressed on the cell surface of leukocytes and mediates their homing to inflammatory sites and peripheral lymph nodes by enabling rolling along the venular wall. CD62L is also involved in activation-induced neutrophil aggregation. Activation-dependent CD62L shedding, however, counteracts neutrophil rolling. CD62L has also signaling roles including enhance of chemokine receptor expression. Similarly to CD62P, the major ligand of CD62L is PSGL-1 (P-selectin glycoprotein ligand-1).
Specifications
Specifications
CD62L
Monoclonal
FITC
SELL
Liquid
SELL
Human CD62L
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
Dreg-56
PBS with BSA and 0.1% sodium azide
P14151
SELL, LAM-1, LAM1, LECAM1, LNHR, LSEL, LYAM1, Leu-8, Lyam-1, PLNHR, TQ1, hLHRc
Mouse
IgG1, kappa
0.5 mL
4° C, store in dark
Primary
6402