Learn More
CD62L Mouse anti-Human, PE, Clone: Dreg-56, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD62L04
Catalog No. 50-112-1448
Add to cart
Description
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD62L (L-selectin) is an adhesion glycoprotein that is constitutively expressed on the cell surface of leukocytes and mediates their homing to inflammatory sites and peripheral lymph nodes by enabling rolling along the venular wall. CD62L is also involved in activation-induced neutrophil aggregation. Activation-dependent CD62L shedding, however, counteracts neutrophil rolling. CD62L has also signaling roles including enhance of chemokine receptor expression. Similarly to CD62P, the major ligand of CD62L is PSGL-1 (P-selectin glycoprotein ligand-1).
Specifications
Specifications
CD62L
Monoclonal
PE
SELL
Liquid
SELL
Human CD62L
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
Dreg-56
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide
P14151
SELL, LAM-1, LAM1, LECAM1, LNHR, LSEL, LYAM1, Leu-8, Lyam-1, PLNHR, TQ1, hLHRc
Mouse
IgG1, kappa
0.5 mL
4° C, store in dark
Primary
6402