Learn More
CD62P (P-Selectin) Mouse anti-Human, BB700, Clone: AK-4, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566565
Catalog No. BDB566565
Add to cart
Description
Description
CD62P (P-Selectin) Monoclonal antibody specifically detects CD62P (P-Selectin) in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD62P (P-Selectin)
AK-4 (also known as AK4 or AK 4)
Conjugated, Purified
IgG1, κ
RUO
Primary
CD62P (P-Selectin)
Flow Cytometry
BB700
Mouse
100 Tests
Refrigerate (2 to 8°C)
Monoclonal
Human