Learn More
CD63 Antibody (OTI5E5), Biotin, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270380B
Catalog No. NBP270380B
Add to Cart
Description
Description
CD63 Monoclonal specifically detects CD63 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
CD63
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
CD63
Full length recombinant protein of human CD63 (NP_001771) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Western Blot, Immunohistochemistry
OTI5E5
Western Blot, Immunohistochemistry
CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin
Mouse
Affinity Purified
Autophagy, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis
967.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions